Learn More
Biolegend Pacific Blue™ anti-mouse IL-17A
Supplier:  Biolegend 506918
Encompass_Preferred
Pacific Blue™ anti-mouse IL-17A [TC11-18H10.1]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: ICFC; Size: 100 ug
Catalog No. 50-170-006