Learn More
Biolegend Pacific Blue™ anti-mouse Ly-49A
Supplier:  Biolegend 116810
Encompass_Preferred
Pacific Blue™ anti-mouse Ly-49A [YE1/48.10.6]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-163-699