Learn More
Biolegend Pacific Blue™ anti-mouse/rat CD29
Supplier:  Biolegend 102224
Encompass_Preferred
Pacific Blue™ anti-mouse/rat CD29 [HMβ1-1]; Isotype: Armenian Hamster IgG; Reactivity: Mouse, Rat; Apps: FC; Size: 100 ug
Catalog No. 50-162-669