Learn More
Biolegend Pacific Blue™ anti-mouse Siglec H
Supplier:  Biolegend 129609
Encompass_Preferred
Pacific Blue™ anti-mouse Siglec H [551]; Isotype: Rat IgG1; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-164-518