Learn More
AllCells LLC Peripheral Blood, Cryopreserved, CD4+ Helper T Cells, Negatively Selected, 25M
Catalog No. 50156846
Encompass
CD4+ Helper T cells are negatively selected from peripheral blood leukapheresis using immunomagnetic microbeads targeting non-helper T cells. CD4+ Helper T cells are shipped in a 2mL cryovial. Purity is determined prior to cryopreservation and is ≥ 85%. Viability is ≥ 80% post-thaw.
Catalog No. 50-156-846 Supplier AllCells LLC Supplier No. LP, CR, CD4+, NS, 25M
CD4+ Helper T cells are negatively selected from peripheral blood leukapheresis using immunomagnetic microbeads targeting non-helper T cells. CD4+ Helper T cells are shipped in a 2mL cryovial. Purity is determined prior to cryopreservation and is ≥ 85%. Viability is ≥ 80% post-thaw.