Learn More
AllCells LLC Peripheral Blood, Cryopreserved, CD8+ Cytotoxic T Cells, Negatively Selected, 10M
Catalog No. 50156850
Encompass
CD8+ Cytotoxic T cells are negatively selected from peripheral blood leukapheresis using immunomagnetic microbeads targeting non-cytotoxic T cells. CD8+ Cytotoxic T cells are shipped in a 2mL cryovial. Purity is determined prior to cryopreservation and is ≥ 85%. Viability is ≥ 80% post-thaw.
Catalog No. 50-156-850 Supplier AllCells LLC Supplier No. LP, CR, CD8+, NS, 10M
CD8+ Cytotoxic T cells are negatively selected from peripheral blood leukapheresis using immunomagnetic microbeads targeting non-cytotoxic T cells. CD8+ Cytotoxic T cells are shipped in a 2mL cryovial. Purity is determined prior to cryopreservation and is ≥ 85%. Viability is ≥ 80% post-thaw.