Learn More
Charles River Labs Cell Solutions PB CD8 CELLS- NEG SEL CRYO 10M
Catalog No. 50970353
Encompass
For Research Use Only. Human Normal Peripheral Blood CD8+ Cytotoxic T Cells - Cryopreserved, 10 million cells/vial. Isolated using NEGATIVE immunomagnetic cell separation procedures.
Catalog No. 50-970-353 Supplier Charles River Labs Cell Solutions Supplier No. PB08NC2
For Research Use Only. Human Normal Peripheral Blood CD8+ Cytotoxic T Cells - Cryopreserved, 10 million cells/vial. Isolated using NEGATIVE immunomagnetic cell separation procedures.