Learn More
Charles River Labs Cell Solutions PB CD8 CELLS- POS SEL CRYO 10M
Catalog No. 50970350
Encompass
For Research Use Only. Human Normal Peripheral Blood CD8+ Cytotoxic T Cells - Cryopreserved, 10 million cells/vial. Isolated using POSITIVE immunomagnetic cell separation procedures.
Catalog No. 50-970-350 Supplier Charles River Labs Cell Solutions Supplier No. PB08C2
For Research Use Only. Human Normal Peripheral Blood CD8+ Cytotoxic T Cells - Cryopreserved, 10 million cells/vial. Isolated using POSITIVE immunomagnetic cell separation procedures.