Learn More
System Biosciences LLC PB-CMV-MCS-EF1α-Puro PiggyBac cDNA Cloning and Expression VectorPB-CMV-MCS-EF1α-Puro cDNA cloning and expression vector = 10 µg
SDP
Catalog No. NC1271869
Encompass_Preferred
Easily deliver your gene-of-interest using PiggyBac – this vector includes a puromycin resistance gene and drives your cDNA with the strong CMV promoter. Get easy, consistent transgenesis Consistent and easy-to-use, SBI’s PiggyBac Transposon System includes cloning and expression vectors that come with a range of markers as well as both constitutive and inducible promoters. The PB-CMV-MCS-EF1α-Puro PiggyBac cDNA Cloning and Expression Vector (Cat.# PB510B-1) drives expression of your gene-of-interest from the strong CMV promoter and expression of a puromycin resistance marker from the EF1α promoter.
Catalog No. NC1271869 Supplier System Biosciences LLC Supplier No. PB510B1
Easily deliver your gene-of-interest using PiggyBac – this vector includes a puromycin resistance gene and drives your cDNA with the strong CMV promoter. Get easy, consistent transgenesis Consistent and easy-to-use, SBI’s PiggyBac Transposon System includes cloning and expression vectors that come with a range of markers as well as both constitutive and inducible promoters. The PB-CMV-MCS-EF1α-Puro PiggyBac cDNA Cloning and Expression Vector (Cat.# PB510B-1) drives expression of your gene-of-interest from the strong CMV promoter and expression of a puromycin resistance marker from the EF1α promoter.