Learn More
System Biosciences LLC PB-CMV-MCS-EF1α-RedPuro PiggyBac cDNA Cloning and Expression VectorPB-CMV-MCS-EF1α-RedPuro cDNA Cloning and Expression Vector = 10 µg
SDP
Catalog No. NC2399456
Encompass_Preferred
Easily deliver your gene-of-interest using PiggyBac – this vector includes both RFP and puro reporters and drives your cDNA with the strong CMV promoter. Consistent and easy-to-use, SBI’s PiggyBac Transposon System includes cloning and expression vectors that come with a range of markers as well as both constitutive and inducible promoters. The PB-CMV-MCS-EF1α-RedPuro PiggyBac cDNA Cloning and Expression Vector (Cat.# PB514B-1) drives expression of your gene-of-interest from the strong CMV promoter and expression of both an RFP reporter and puromycin resistance from the EF1α promoter.
Catalog No. NC2399456 Supplier System Biosciences LLC Supplier No. PB514B2
Easily deliver your gene-of-interest using PiggyBac – this vector includes both RFP and puro reporters and drives your cDNA with the strong CMV promoter. Consistent and easy-to-use, SBI’s PiggyBac Transposon System includes cloning and expression vectors that come with a range of markers as well as both constitutive and inducible promoters. The PB-CMV-MCS-EF1α-RedPuro PiggyBac cDNA Cloning and Expression Vector (Cat.# PB514B-1) drives expression of your gene-of-interest from the strong CMV promoter and expression of both an RFP reporter and puromycin resistance from the EF1α promoter.