Learn More
Charles River Labs Cell Solutions Peripheral Blood Pan Monocytes (CD14+ and/or CD16+) (Negative Selection), 10 Million cells, Cryopreserved
Supplier:  Charles River Labs Cell Solutions PB1416NC1
Encompass
Pan monocytes (which include classical, intermediate, and non-classical monocytes) are responsible for phagocytosis of foreign substances in the body and are capable of killing infected host cells via antibody-mediated cellular cytotoxicity. Under the right culture conditions, monocytes can differentiate into dendritic cells, Langerhans cells, or macrophages. Pan monocytes are isolated from mononuclear cells by negative selection using immunomagnetic cell separation procedures.
Catalog No. 50-225-6296
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov