Learn More
Charles River Labs Cell Solutions PB DENDRITIC CELLS CRYO 2M
Catalog No. 50970337
Encompass
For Research Use Only. Human Normal Peripheral Blood Dendritic Cells - Cryopreserved, 2 million cells/vial. Derived from purified CD14 monocytes.CD14 monocytes are isolated from PBMCs and are cultured in the presence of GM-CSF and IL-4 for 7 days.
Catalog No. 50-970-337 Supplier Charles River Labs Cell Solutions Supplier No. PB010C1
For Research Use Only. Human Normal Peripheral Blood Dendritic Cells - Cryopreserved, 2 million cells/vial. Derived from purified CD14 monocytes.CD14 monocytes are isolated from PBMCs and are cultured in the presence of GM-CSF and IL-4 for 7 days.