Learn More
AllCells LLC Peripheral Blood, Fresh, Mononuclear Cells, 2 Billion
Catalog No. 502067632
Encompass
Peripheral blood cells collected by apheresis are treated with ammonium chloride to remove red blood cells (RBCs) and washed to remove platelets. Viability is 90%. Our peripheral blood MNC products (PBMCs) are composed of, on average, 50% T cells, 20% monocytes, 10% B cells, 7% NK cells, and 3% granulocytes. Actual cell type percentages vary from collection to collection.
Catalog No. 50-206-7632 Supplier AllCells LLC Supplier No. LP, FR, MNC, 2B
Peripheral blood cells collected by apheresis are treated with ammonium chloride to remove red blood cells (RBCs) and washed to remove platelets. Viability is 90%. Our peripheral blood MNC products (PBMCs) are composed of, on average, 50% T cells, 20% monocytes, 10% B cells, 7% NK cells, and 3% granulocytes. Actual cell type percentages vary from collection to collection.