Learn More
AllCells LLC Peripheral Blood, Fresh, Mononuclear Cells, 4 Billion
Catalog No. 502067633
Encompass
Peripheral blood cells collected by apheresis are treated with ammonium chloride to remove red blood cells (RBCs) and washed to remove platelets. Viability is 90%. Our peripheral blood MNC products (PBMCs) are composed of, on average, 50% T cells, 20% monocytes, 10% B cells, 7% NK cells, and 3% granulocytes. Actual cell type percentages vary from collection to collection.
Catalog No. 50-206-7633 Supplier AllCells LLC Supplier No. LP, FR, MNC, 4B
Add to Cart
Add to Cart
Request a Quote
Peripheral blood cells collected by apheresis are treated with ammonium chloride to remove red blood cells (RBCs) and washed to remove platelets. Viability is 90%. Our peripheral blood MNC products (PBMCs) are composed of, on average, 50% T cells, 20% monocytes, 10% B cells, 7% NK cells, and 3% granulocytes. Actual cell type percentages vary from collection to collection.