Learn More
Charles River Labs Cell Solutions PB LEUKOPAK - FULL BAG 12B
Catalog No. 50970317
Encompass
LeukoPaks are collected in HemaCare's FDA-registered collection centers from IRB consented healthy human volunteer donors by leukapheresis using the COBE� Spectra or Spectra Optia� Apheresis System MNC collection protocol in ACD-A anticoagulant. Tissue: Peripheral Blood Format: Fresh Disease: Healthy Size: 1 bag
Catalog No. 50-970-317 Supplier Charles River Labs Cell Solutions Supplier No. PB001F1
LeukoPaks are collected in HemaCare's FDA-registered collection centers from IRB consented healthy human volunteer donors by leukapheresis using the COBE� Spectra or Spectra Optia� Apheresis System MNC collection protocol in ACD-A anticoagulant. Tissue: Peripheral Blood Format: Fresh Disease: Healthy Size: 1 bag