Learn More
Charles River Labs Cell Solutions PB M1 MACROPHAGE CRYO 5M
Catalog No. 50970374
Encompass
For Research Use Only. Human Normal Peripheral Blood M1 Macrophages - Cryopreserved, 5 million cells/vial. Derived from purified Monocytes. CD14 monocytes were positively selected via immunomagnetic beads and were cultured in the presence of GM-CSF for 10 days.
Catalog No. 50-970-374 Supplier Charles River Labs Cell Solutions Supplier No. PBM1CMON2
For Research Use Only. Human Normal Peripheral Blood M1 Macrophages - Cryopreserved, 5 million cells/vial. Derived from purified Monocytes. CD14 monocytes were positively selected via immunomagnetic beads and were cultured in the presence of GM-CSF for 10 days.