Learn More
AllCells LLC Peripheral Blood, Cryopreserved, Mononuclear Cells (PBMC), 100M
Supplier:  AllCells LLC LP, CR, MNC, 100M
Encompass
Peripheral blood cells collected by apheresis are treated with ammonium chloride to remove red blood cells (RBCs) and washed to remove platelets. Viability is 90%. Our peripheral blood MNC products (PBMCs) are composed of, on average, 50% T cells, 20% monocytes, 10% B cells, 7% NK cells, and 3% granulocytes. Actual cell type percentages vary from collection to collection.
Catalog No. 50-156-486