Learn More
Charles River Labs Cell Solutions PB NK CELLS- NEG SEL CRYO 5M
Catalog No. 50970371
Encompass
For Research Use Only. Human Normal Peripheral Blood CD56+ Natural Killer (NK) Cells - Cryopreserved, 5 million cells/vial. Isolated using NEGATIVE immunomagnetic cell separation procedures.
Catalog No. 50-970-371 Supplier Charles River Labs Cell Solutions Supplier No. PB56NC1
For Research Use Only. Human Normal Peripheral Blood CD56+ Natural Killer (NK) Cells - Cryopreserved, 5 million cells/vial. Isolated using NEGATIVE immunomagnetic cell separation procedures.