Learn More
Medchemexpress LLC PBA-1105 TFA | 99.6% | 754.83 | 5 MG
SDP
Catalog No. 5000391483
Encompass_Preferred
PBA-1105 TFA is an autophagy-targeting chimera (AUTOTAC) that induces p62 self-oligomerization. It selectively binds to exposed hydrophobic regions of misfolded proteins, facilitating their degradation via the autophagic pathway. This compound increases the autophagic flux of Ub-conjugated aggregates and induces autophagic degradation of misfolded proteins and aggregates at nanomolar concentrations. It is for research use only.
  • Induces p62 self-oligomerization.
  • Selectively binds to exposed hydrophobic regions of misfolded proteins.
  • Facilitates degradation via the autophagic pathway.
  • Increases the autophagic flux of Ub-conjugated aggregates.
  • Induces autophagic degradation of mutant tau and misfolded proteins.
Catalog No. 50-003-91483 Supplier Medchemexpress LLC Supplier No. HY161743A5MG
PBA-1105 TFA is an autophagy-targeting chimera (AUTOTAC) that induces p62 self-oligomerization. It selectively binds to exposed hydrophobic regions of misfolded proteins, facilitating their degradation via the autophagic pathway. This compound increases the autophagic flux of Ub-conjugated aggregates and induces autophagic degradation of misfolded proteins and aggregates at nanomolar concentrations. It is for research use only.
  • Induces p62 self-oligomerization.
  • Selectively binds to exposed hydrophobic regions of misfolded proteins.
  • Facilitates degradation via the autophagic pathway.
  • Increases the autophagic flux of Ub-conjugated aggregates.
  • Induces autophagic degradation of mutant tau and misfolded proteins.