Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pbd dimer | 1222490-34-7 | MFCD27952508 | 98.5% | 725.79 g/mol | C42H39N5O7 | 10 MG
SDP

Catalog No. 5000213519
Encompass_Preferred

SGD-1882 is a cytotoxic pyrrolobenzodiazepine (PBD) dimer that functions as a DNA minor-groove crosslinking agent and is utilized as a payload in antibody-drug conjugate (ADC) research and development.

  • Cytotoxic pyrrolobenzodiazepine dimer for antibody-drug conjugate payload applications.
  • DNA minor-groove crosslinking mechanism.
  • High purity suitable for conjugation and biological studies.
  • Solid form with light yellow to yellow appearance for handling.
  • Molecular formula C42H39N5O7; molecular weight 725.79 g/mol.
  • Recommended storage: powder at -20°C for long-term stability.

Catalog No. 50-002-13519 Supplier Medchemexpress LLC Supplier No. HY10112710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SGD-1882 is a cytotoxic pyrrolobenzodiazepine (PBD) dimer that functions as a DNA minor-groove crosslinking agent and is utilized as a payload in antibody-drug conjugate (ADC) research and development.

  • Cytotoxic pyrrolobenzodiazepine dimer for antibody-drug conjugate payload applications.
  • DNA minor-groove crosslinking mechanism.
  • High purity suitable for conjugation and biological studies.
  • Solid form with light yellow to yellow appearance for handling.
  • Molecular formula C42H39N5O7; molecular weight 725.79 g/mol.
  • Recommended storage: powder at -20°C for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.