Learn More
Medchemexpress LLC PBD dimer-2 | 145325-57-1 | 98.0% | 560.64 g/mol | C31H36N4O6 | 1 ML
SDP
Catalog No. 5000203587
Encompass_Preferred
PBD dimer-2 is a C8-linked pyrrolobenzodiazepine dimer used as a DNA minor-groove crosslinking agent and as a payload in antibody-drug conjugate research. It is supplied as a solid and as a 10 mM solution in DMSO.
  • C8-linked pyrrolobenzodiazepine dimer suitable for DNA cross-linking studies.
  • High purity (≈98%) for research applications.
  • Available as 10 mM in DMSO (1 ML) or as solid in multiple mg sizes.
  • Soluble in DMSO (≈130 mg/mL); ultrasonic assistance may be required.
  • Store protected from light at -20°C; in solvent store at -80°C for long-term stability.
Catalog No. 50-002-03587 Supplier Medchemexpress LLC Supplier No. HY14842410MM 1ML
PBD dimer-2 is a C8-linked pyrrolobenzodiazepine dimer used as a DNA minor-groove crosslinking agent and as a payload in antibody-drug conjugate research. It is supplied as a solid and as a 10 mM solution in DMSO.
  • C8-linked pyrrolobenzodiazepine dimer suitable for DNA cross-linking studies.
  • High purity (≈98%) for research applications.
  • Available as 10 mM in DMSO (1 ML) or as solid in multiple mg sizes.
  • Soluble in DMSO (≈130 mg/mL); ultrasonic assistance may be required.
  • Store protected from light at -20°C; in solvent store at -80°C for long-term stability.