Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PBD-monoamide | 2093165-00-3 | 96.4% | 600.66 g/mol | C33H36N4O7 | 10 MG
SDP

Catalog No. 5000202433
Encompass_Preferred

PBD-monoamide is a modified pyrrolobenzodiazepine (PBD) dimer used as an antibody-drug conjugate (ADC) cytotoxin intermediate for research applications. The material is supplied in a small-mass vial and requires protection from light and inert storage. Reported analytical data include purity and molecular properties.

  • Used as an antibody-drug conjugate (ADC) cytotoxin intermediate
  • Reported purity 96.4%
  • Molecular formula C33H36N4O7 and molecular weight 600.66 g/mol
  • Supplied in 10 MG vial for small-scale laboratory use
  • Storage: protect from light; solid at 4°C, in solvent -80°C (6 months) or -20°C (1 month)

Catalog No. 50-002-02433 Supplier Medchemexpress LLC Supplier No. HY16125210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PBD-monoamide is a modified pyrrolobenzodiazepine (PBD) dimer used as an antibody-drug conjugate (ADC) cytotoxin intermediate for research applications. The material is supplied in a small-mass vial and requires protection from light and inert storage. Reported analytical data include purity and molecular properties.

  • Used as an antibody-drug conjugate (ADC) cytotoxin intermediate
  • Reported purity 96.4%
  • Molecular formula C33H36N4O7 and molecular weight 600.66 g/mol
  • Supplied in 10 MG vial for small-scale laboratory use
  • Storage: protect from light; solid at 4°C, in solvent -80°C (6 months) or -20°C (1 month)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.