Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PBD-monoamide | 2093165-00-3 | 96.4% | 600.66 g·mol⁻¹ | C33H36N4O7 | 50 MG
SDP

Catalog No. 5000202800
Encompass_Preferred

PBD-monoamide is a pyrrolobenzodiazepine (PBD)-based antibody-drug conjugate (ADC) cytotoxin with DNA-binding activity, used as an ADC payload and for synthesis of experimental HER2 ADCs. Intended for research use only; not for human use.

  • ADC cytotoxin with DNA-binding activity
  • Used for synthesis of HER2-directed ADCs
  • Physical state: solid with high purity
  • Key identifiers: CAS 2093165-00-3; formula C33H36N4O7; Mw 600.66 g·mol⁻¹
  • Purity approximately 96.4%
  • Storage: protect from light, store under inert gas; keep at 4°C for solid

Catalog No. 50-002-02800 Supplier Medchemexpress LLC Supplier No. HY16125250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

PBD-monoamide is a pyrrolobenzodiazepine (PBD)-based antibody-drug conjugate (ADC) cytotoxin with DNA-binding activity, used as an ADC payload and for synthesis of experimental HER2 ADCs. Intended for research use only; not for human use.

  • ADC cytotoxin with DNA-binding activity
  • Used for synthesis of HER2-directed ADCs
  • Physical state: solid with high purity
  • Key identifiers: CAS 2093165-00-3; formula C33H36N4O7; Mw 600.66 g·mol⁻¹
  • Purity approximately 96.4%
  • Storage: protect from light, store under inert gas; keep at 4°C for solid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.