Learn More
Cell Signaling Technology PBEF/NAMPT Monoclonal Antibody, Rabbit, 100 μl
SDP
Catalog No. 502732601
Encompass
PBEF/NAMPT (D1K6D) Rabbit Monoclonal Antibody is designed to recognize endogenous levels of total PBEF/NAMPT protein. This monoclonal antibody is generated by immunizing animals with a synthetic peptide corresponding to residues surrounding Gly122 of the human PBEF/NAMPT protein. NAMPT catalyzes the rate-limiting step in the NAD biosynthesis pathway starting from nicotinamide and is critical for glucose-stimulated insulin secretion in pancreatic beta cells.
  • Validated for western blotting, immunoprecipitation, and immunohistochemistry (paraffin) applications.
  • Species reactivity is limited to human.
  • Supplied in a formulation containing 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/ml BSA, and 50% glycerol.
  • Recommended for research use only; not for use in diagnostic procedures.
Catalog No. 50-273-2601 Supplier Cell Signaling Technology Supplier No. 61122S
PBEF/NAMPT (D1K6D) Rabbit Monoclonal Antibody is designed to recognize endogenous levels of total PBEF/NAMPT protein. This monoclonal antibody is generated by immunizing animals with a synthetic peptide corresponding to residues surrounding Gly122 of the human PBEF/NAMPT protein. NAMPT catalyzes the rate-limiting step in the NAD biosynthesis pathway starting from nicotinamide and is critical for glucose-stimulated insulin secretion in pancreatic beta cells.
  • Validated for western blotting, immunoprecipitation, and immunohistochemistry (paraffin) applications.
  • Species reactivity is limited to human.
  • Supplied in a formulation containing 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/ml BSA, and 50% glycerol.
  • Recommended for research use only; not for use in diagnostic procedures.