Learn More
Enzo Life Sciences Visfatin/Nampt monoclonal antibody (P4D5AT) (50µg)
SDP
Supplier:  Enzo Life Sciences ALX804922C050
Encompass_Preferred
Clone: P4D5AT. Isotype: IgG1. Immunogen: Recombinant human Visfatin (Nampt/PBEF) (aa 1-491). Species reactivity: Human. Applications: ELISA, WB. Purity: Protein G-affinity purified. Formulation: Liquid. In PBS, pH 7.4, containing 0.1% sodium azide.
Catalog No. 50-201-0025