Learn More
Aviva Systems Biology Corp PBEF1 Antibody - C-terminal region (ARP42254_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42254_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against PBEF1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human PBEF1. Alternate names: 1110035O14Rik, DKFZP666B131, MGC117256, NAMPT, PBEF, VF, PBEF1, VISFATINApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53330