Learn More
Medchemexpress LLC HY-100775 5mg Medchemexpress, PBI-4050 (sodium salt) CAS:1254472-97-3 Purity:>98%
SDP
Catalog No. 501931945
Encompass_Preferred
Medchemexpress, HY-100775 5mg PBI-4050 (sodium salt) CAS:1254472-97-3 PBI-4050 sodium salt acts as an agonist for GPR40 and as an antagonist or inverse agonist for GPR84 . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1945 Supplier Medchemexpress LLC Supplier No. HY1007755MG
Medchemexpress, HY-100775 5mg PBI-4050 (sodium salt) CAS:1254472-97-3 PBI-4050 sodium salt acts as an agonist for GPR40 and as an antagonist or inverse agonist for GPR84 . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.