Learn More
TARGETMOL CHEMICALS INC PBIT 10MG
SDP
Catalog No. 502577338
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PBIT inhibits JARID1B histone demethylase (IC50 of about 3 uM). PBIT also inhibits JARID1A and JARID1C (IC50s of 6 uM and 4.9 uM respectively). PBIT is a specific inhibitor of the Jumonji AT-rich Interactive Domain 1 (JARID1) enzymes . purity: 99%
Catalog No. 50-257-7338 Supplier TARGETMOL CHEMICALS INC Supplier No. T1643510MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PBIT inhibits JARID1B histone demethylase (IC50 of about 3 uM). PBIT also inhibits JARID1A and JARID1C (IC50s of 6 uM and 4.9 uM respectively). PBIT is a specific inhibitor of the Jumonji AT-rich Interactive Domain 1 (JARID1) enzymes . purity: 99%