Learn More
Aviva Systems Biology Corp PBLD Antibody - N-terminal region (ARP56239_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56239_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PBLD. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PBLD. Alternate names: FLJ14767, FLJ35507, MAWBP, MAWDBPApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Yeast
Catalog No. 50-014-61778