Learn More
Agilent Technologies PBLUESCRIPT2KS(+)PHAGEMIDKIT
Catalog No. 50125247
Encompass
pBluescript II KS( + ) Phagemid Kit 1 kit f1 origin in ( + ) orientation Kpn--> Sac polylinker orientation Contains: 20microgram pBluescript II KS( + ) phagemid vector Host Strain: XL1-Blue MRF
Catalog No. 50-125-247 Supplier Agilent Technologies Supplier No. 212207
pBluescript II KS( + ) Phagemid Kit 1 kit f1 origin in ( + ) orientation Kpn--> Sac polylinker orientation Contains: 20microgram pBluescript II KS( + ) phagemid vector Host Strain: XL1-Blue MRF