Learn More
Agilent Technologies PBLUESCRIPT2SK(+)PHAGEMIDKIT
Catalog No. 50125245
Encompass
pBluescript II SK( + ) Phagemid Kit 1 kit f1 origin in ( + ) orientation Sac--> Kpn polylinker orientation Contains: 20microgram pBluescript II SK( + ) phagemid vector Host Strain: XL1-Blue MRF
Catalog No. 50-125-245 Supplier Agilent Technologies Supplier No. 212205
pBluescript II SK( + ) Phagemid Kit 1 kit f1 origin in ( + ) orientation Sac--> Kpn polylinker orientation Contains: 20microgram pBluescript II SK( + ) phagemid vector Host Strain: XL1-Blue MRF