Learn More
Charles River Labs Cell Solutions PBMC CRYO 100M
Catalog No. 50970332
Encompass
For Research Use Only. Human Normal Peripheral Blood Mononuclear Cells (PBMC) - Cryopreserved, 100 million cells/vial. Mononuclear cells (PBMCs) are purified using density gradient centrifugation techniques.
Catalog No. 50-970-332 Supplier Charles River Labs Cell Solutions Supplier No. PB009C3
For Research Use Only. Human Normal Peripheral Blood Mononuclear Cells (PBMC) - Cryopreserved, 100 million cells/vial. Mononuclear cells (PBMCs) are purified using density gradient centrifugation techniques.