Learn More
Charles River Labs Cell Solutions PBMC CRYO 10M
Catalog No. 50970329
Encompass
For Research Use Only. Human Normal Peripheral Blood Mononuclear Cells (PBMC) - Cryopreserved, 10 million cells/vial. Mononuclear cells (PBMCs) are purified using density gradient centrifugation techniques.
Catalog No. 50-970-329 Supplier Charles River Labs Cell Solutions Supplier No. PB009C1
For Research Use Only. Human Normal Peripheral Blood Mononuclear Cells (PBMC) - Cryopreserved, 10 million cells/vial. Mononuclear cells (PBMCs) are purified using density gradient centrifugation techniques.