Learn More
Charles River Labs Cell Solutions PBMC CRYO 25M
Catalog No. 50970330
Encompass
For Research Use Only. Human Normal Peripheral Blood Mononuclear Cells (PBMC) - Cryopreserved, 25 million cells/vial. Mononuclear cells (PBMCs) are purified using density gradient centrifugation techniques.
Catalog No. 50-970-330 Supplier Charles River Labs Cell Solutions Supplier No. PB009C2
For Research Use Only. Human Normal Peripheral Blood Mononuclear Cells (PBMC) - Cryopreserved, 25 million cells/vial. Mononuclear cells (PBMCs) are purified using density gradient centrifugation techniques.