Learn More
iXCells Biotechnologies Peripheral Blood Mononuclear Cells (PBMCs), 10M
Catalog No. 501077993
Encompass_Preferred
Peripheral Blood Mononuclear Cells (PBMCs) is any blood cell with a single nucleus. PBMCs consist of lymphocytes (T, B and NK cells), monocytes and dendritic cells. iXCells Biotechnologies offers PBMCs from the human peripheral blood of individual healthy donors. These PBMCs are purified from peripheral blood using ficoll gradient centrifugation.
Catalog No. 50-107-7993 Supplier iXCells Biotechnologies Supplier No. 10HU003CR10M
Peripheral Blood Mononuclear Cells (PBMCs) is any blood cell with a single nucleus. PBMCs consist of lymphocytes (T, B and NK cells), monocytes and dendritic cells. iXCells Biotechnologies offers PBMCs from the human peripheral blood of individual healthy donors. These PBMCs are purified from peripheral blood using ficoll gradient centrifugation.