Learn More
Enzo Life Sciences PBN, (1g), CAS Number: 3376-24-7
SDP
Catalog No. NC0958233
Encompass_Preferred
Spin trapping reagent. Cell permeable spin trap for both in vivo and in vitro studies. Alternative Name: N-tert-Butyl-alpha-phenylnitrone. Formula: C11H15NO. MW: 177.3. CAS Number: 3376-24-7. MI: 14: 7056. Purity: ≥98% (HPLC). Appearance: White to off-white crystalline solid. Solubility: Soluble in DMSO or 100% ethanol; slightly soluble in water. Long Term Storage: -20°C
Catalog No. NC0958233 Supplier Enzo Life Sciences Supplier No. ALX430082G001
Spin trapping reagent. Cell permeable spin trap for both in vivo and in vitro studies. Alternative Name: N-tert-Butyl-alpha-phenylnitrone. Formula: C11H15NO. MW: 177.3. CAS Number: 3376-24-7. MI: 14: 7056. Purity: ≥98% (HPLC). Appearance: White to off-white crystalline solid. Solubility: Soluble in DMSO or 100% ethanol; slightly soluble in water. Long Term Storage: -20°C