Learn More
Medchemexpress LLC HY-U00446 5mg Medchemexpress, PBOX 6 CAS:290814-68-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYU004465MG
Encompass_Preferred
Medchemexpress, HY-U00446 5mg PBOX 6 CAS:290814-68-5 PBOX 6 is a pyrrolo-1,5-benzoxazepine (PBOX) compound, acts as a microtubule -depolymerizing agent and an apoptotic agent. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4454