Learn More
Medchemexpress LLC PBP10 TFA | 794466-43-6 | 98.22% | 1826.07 | 5 MG
SDP
Catalog No. 5000424599
Encompass_Preferred
PBP10 TFA is a cell-permeable and selective gelsolin-derived peptide inhibitor of formyl peptide receptor 2 (FPR2) over FPR1. This 10-amino acid peptide is N-terminally conjugated with rhodamine. It demonstrates bactericidal activity against both gram-positive and gram-negative bacteria and limits microbial-induced inflammatory effects. The compound is for research use only.
  • Selective inhibitor of formyl peptide receptor 2 (FPR2)
  • Cell-permeable gelsolin-derived peptide
  • 10-amino acid peptide with rhodamine conjugated at its N-terminus
  • Exerts bactericidal activity against gram-positive and gram-negative bacteria
  • Limits microbial-induced inflammatory effects
Catalog No. 50-004-24599 Supplier Medchemexpress LLC Supplier No. HYP1116A5MG
PBP10 TFA is a cell-permeable and selective gelsolin-derived peptide inhibitor of formyl peptide receptor 2 (FPR2) over FPR1. This 10-amino acid peptide is N-terminally conjugated with rhodamine. It demonstrates bactericidal activity against both gram-positive and gram-negative bacteria and limits microbial-induced inflammatory effects. The compound is for research use only.
  • Selective inhibitor of formyl peptide receptor 2 (FPR2)
  • Cell-permeable gelsolin-derived peptide
  • 10-amino acid peptide with rhodamine conjugated at its N-terminus
  • Exerts bactericidal activity against gram-positive and gram-negative bacteria
  • Limits microbial-induced inflammatory effects