Learn More
Medchemexpress LLC PBP10 TFA | 98.2% | 1826.07 | 1 MG
SDP
Catalog No. 5000423427
Encompass_Preferred
PBP10 is a cell-permeable and selective gelsolin-derived peptide inhibitor of formyl peptide receptor 2 (FPR2) over FPR1. This 10-AA peptide, with rhodamine conjugated at its N-terminus, exhibits bactericidal activity against gram-positive and gram-negative bacteria and mitigates microbial-induced inflammatory effects.
  • Cell-permeable
  • Selective gelsolin-derived peptide inhibitor of FPR2 over FPR1
  • 10-AA peptide with rhodamine conjugated at its N-terminus
  • Exerts bactericidal activity against gram-positive and gram-negative bacteria
  • Limits microbial-induced inflammatory effects
Catalog No. 50-004-23427 Supplier Medchemexpress LLC Supplier No. HYP1116A1MG
PBP10 is a cell-permeable and selective gelsolin-derived peptide inhibitor of formyl peptide receptor 2 (FPR2) over FPR1. This 10-AA peptide, with rhodamine conjugated at its N-terminus, exhibits bactericidal activity against gram-positive and gram-negative bacteria and mitigates microbial-induced inflammatory effects.
  • Cell-permeable
  • Selective gelsolin-derived peptide inhibitor of FPR2 over FPR1
  • 10-AA peptide with rhodamine conjugated at its N-terminus
  • Exerts bactericidal activity against gram-positive and gram-negative bacteria
  • Limits microbial-induced inflammatory effects