Learn More
New England Biolabs, Inc. pBR322 DNA-MspI Digest – 0.05 ml
SDP
Catalog No. 50811767
Encompass
The MspI digest of pBR322 DNA yields 26 fragments. Suitable for use as molecular weight standards for agarose gel electrophoresis. Comes supplied with 1 vial of Gel Loading Dye, Purple (6X), no SDS.
Catalog No. 50-811-767 Supplier New England Biolabs, Inc. Supplier No. N3032S
The MspI digest of pBR322 DNA yields 26 fragments. Suitable for use as molecular weight standards for agarose gel electrophoresis. Comes supplied with 1 vial of Gel Loading Dye, Purple (6X), no SDS.