Learn More
Sigma Aldrich Fine Chemicals Biosciences pBR322 Hae III Digest for1MG
Catalog No. 502907082
Encompass
Sigmas pBR322 Hae III Digest contains 22 fragments from 8 - 587 bp. Ten ul of the ladder should be diluted in gel loading buffer and then loaded in a single lane on an agarose or polyacrylamide gel.
Catalog No. 50-290-7082 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. D9655.1MG
Sigmas pBR322 Hae III Digest contains 22 fragments from 8 - 587 bp. Ten ul of the ladder should be diluted in gel loading buffer and then loaded in a single lane on an agarose or polyacrylamide gel.