Learn More
New England Biolabs, Inc. pBR322 Vector – 250 µg
SDP
Catalog No. 50811768
Encompass
pBR322 DNA is a commonly used plasmid cloning vector in E. coli and is a double-stranded circle 4,361* base pairs in length. pBR322 contains the genes for resistance to ampicillin and tetracycline, and can be amplified with chloramphenicol. The molecular weight is 2.83 x 10^6 daltons.
Catalog No. 50-811-768 Supplier New England Biolabs, Inc. Supplier No. N3033L
pBR322 DNA is a commonly used plasmid cloning vector in E. coli and is a double-stranded circle 4,361* base pairs in length. pBR322 contains the genes for resistance to ampicillin and tetracycline, and can be amplified with chloramphenicol. The molecular weight is 2.83 x 10^6 daltons.