Learn More
Aviva Systems Biology Corp PBRM1 Antibody - N-terminal region (ARP39331_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39331_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PBRM1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human PBRM1. Alternate names: BAF180, MGC156155, MGC156156, PB1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-284-800