Learn More
TARGETMOL CHEMICALS INC PBRM1-BD2-IN-2 5MG
SDP
Catalog No. 502579663
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PBRM1-BD2-IN-2 is a selective and cell-active polybromo-1 (PBRM1) bromodomain inhibitor. PBRM1-BD2-IN-2 has binding affinity and inhibitory activity for PBRM1-BD2 with Kd and IC50 values of 9.3 uM and 1.0 uM respectively. PBRM1-BD2-IN-2 can be used for the research of cancer. purity: 100%
Catalog No. 50-257-9663 Supplier TARGETMOL CHEMICALS INC Supplier No. T601565MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PBRM1-BD2-IN-2 is a selective and cell-active polybromo-1 (PBRM1) bromodomain inhibitor. PBRM1-BD2-IN-2 has binding affinity and inhibitory activity for PBRM1-BD2 with Kd and IC50 values of 9.3 uM and 1.0 uM respectively. PBRM1-BD2-IN-2 can be used for the research of cancer. purity: 100%