Learn More
TARGETMOL CHEMICALS INC PBRM1-BD2-IN-5 5MG
SDP
Catalog No. 502579660
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PBRM1-BD2-IN-5 is a potent PBRM1 Bromodomain inhibitor with Kd values of 1.5 uM and 3.9 uM for PBRM1-BD2 and PBRM1-BD5 respectively and an IC50 value of 0.26 uM for PBRM1-BD2. PBRM1-BD2-IN-5 reduces the binding of full-length PBRM1 within the PBAF complex in cell lysates to acetylated histone peptide. PBRM1-BD2-IN-5 can be used to research anticancer. purity: 99%
Catalog No. 50-257-9660 Supplier TARGETMOL CHEMICALS INC Supplier No. T601595MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PBRM1-BD2-IN-5 is a potent PBRM1 Bromodomain inhibitor with Kd values of 1.5 uM and 3.9 uM for PBRM1-BD2 and PBRM1-BD5 respectively and an IC50 value of 0.26 uM for PBRM1-BD2. PBRM1-BD2-IN-5 reduces the binding of full-length PBRM1 within the PBAF complex in cell lysates to acetylated histone peptide. PBRM1-BD2-IN-5 can be used to research anticancer. purity: 99%