Learn More
FUJIFILM Biosciences PBS 1X-Dulbecco's Phosphate Buffered Saline Solution - Liquid
Catalog No. NC0324574
Encompass
PBS 1X Dulbecco's Phosphate Buffered Saline Solution can be used to provide a buffer system to maintain cell culture media in the physiological pH range, or to irrigate, transport, or dilute fluid while maintaining cell tonicity and viability.
Catalog No. NC0324574 Supplier FUJIFILM Biosciences Supplier No. 92401L
PBS 1X Dulbecco's Phosphate Buffered Saline Solution can be used to provide a buffer system to maintain cell culture media in the physiological pH range, or to irrigate, transport, or dilute fluid while maintaining cell tonicity and viability.