Learn More
Research Products International Corp PBS [Phosphate Buffered Saline] 100 ml Tablets, 100 Tablets per Bottle
SDP
Catalog No. 50488916
Encompass_Preferred
PBS, 100 mL Tablets are pre-weighed tablets used to prepare 100 mL of Phosphate-Buffered Saline, a widely used buffer in biological research. Each tablet contains sodium chloride, sodium phosphate dibasic, and potassium phosphate monobasic, which dissolve in water to form an isotonic, pH-balanced solution. PBS is used in cell culture, immunohistochemistry, protein and nucleic acid research, and diagnostic assays. These tablets ensure consistent buffer preparation, eliminate weighing steps, and save time in lab workflows. They support sample integrity and provide a stable environment for biological reactions.
Catalog No. 50-488-916 Supplier Research Products International Corp Supplier No. P32080100T
PBS, 100 mL Tablets are pre-weighed tablets used to prepare 100 mL of Phosphate-Buffered Saline, a widely used buffer in biological research. Each tablet contains sodium chloride, sodium phosphate dibasic, and potassium phosphate monobasic, which dissolve in water to form an isotonic, pH-balanced solution. PBS is used in cell culture, immunohistochemistry, protein and nucleic acid research, and diagnostic assays. These tablets ensure consistent buffer preparation, eliminate weighing steps, and save time in lab workflows. They support sample integrity and provide a stable environment for biological reactions.