Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ PBS 10X

Catalog No. MP1PBS10X03 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 x 1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP1PBS10X03 5 x 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP1PBS10X03 Supplier MP Biomedicals™ Supplier No. 11PBS10X03
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PBS 10X, Rnase, Dnase, Protease free, Filtered through 0,2μm PTFE.

  • PBS 10 X buffer is ideal for maintaining a constant pH, and since it is isotonic and non-toxic to cells.
  • it can be used in a wide variety of biological applications.
  • PBS 10X is frequently used as a wash and dilution buffer.
  • It is also used to take a reference spectrum when measuring protein adsorption.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phosphate Buffered Saline (PBS)
For Use With (Application) Buffer, Mammalian Cell Culture
Physical Form Liquid
Quantity 5 x 1 L
Grade Molecular Biology
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.