Learn More
Advanced Biomatrix Phosphate Buffered Saline 10X PBS 100 mL
Catalog No. 502255899
Encompass_Preferred
Phosphate Buffered Saline (PBS) 10X  is formulated and prepared to use in conjunction with other products provided by Advanced BioMatrix such as Type I collagen for forming a 3D hydrogel. PBS 10X is provided in a user-friendly packaging for use and storage.  This product is sterile and supplied as a ready to use solution.
Catalog No. 50-225-5899 Supplier Advanced Biomatrix Supplier No. 5076100ML
Phosphate Buffered Saline (PBS) 10X  is formulated and prepared to use in conjunction with other products provided by Advanced BioMatrix such as Type I collagen for forming a 3D hydrogel. PBS 10X is provided in a user-friendly packaging for use and storage.  This product is sterile and supplied as a ready to use solution.