Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quality Biological Inc PBS (10X), pH 7.4, 1000 mL
SDP

Catalog No. 50983226
Encompass_Preferred

PBS is a balanced salt solution used for washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents. Useful in immunological applications, such as Western Blotting.

Without Calcium and Magnesium
Endotoxin and Mycoplasma tested

Storage Condition: 15 °C to 30 °C
Shipping Condition: ambient temperature
Sterile Filtered
Endotoxin and Mycoplasma Tested
Osmolality: 300 ± 55 mOsm/Kg H20
Max Shelf Life: 24 months from date of manufacture

Manufactured under an ISO 9001:2015 certified quality management system.

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Catalog No. 50-983-226 Supplier Quality Biological Inc Supplier No. 119069131
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PBS is a balanced salt solution used for washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents. Useful in immunological applications, such as Western Blotting.

Without Calcium and Magnesium
Endotoxin and Mycoplasma tested

Storage Condition: 15 °C to 30 °C
Shipping Condition: ambient temperature
Sterile Filtered
Endotoxin and Mycoplasma Tested
Osmolality: 300 ± 55 mOsm/Kg H20
Max Shelf Life: 24 months from date of manufacture

Manufactured under an ISO 9001:2015 certified quality management system.

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.